siRNA Details
Gene Symbol: UBAC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036531.1 | 2 | 1657 | |||
NR_036532.1 | 2 | 1383 | ||||
GenBank mRNA | AF339765.1 | - | 1 | 855 | ||
AK097488.1 | - | 2 | 1662 | |||
BC043515.1 | - | 2 | 1338 |
