siRNA Details
Gene Symbol: UBAC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036531.1 | 2 | 447 | |||
NR_036532.1 | 2 | 173 | ||||
GenBank mRNA | AK097488.1 | - | 2 | 452 | ||
BC043515.1 | - | 2 | 128 | |||
DB032212.1 | - | 2 | 448 | |||
DB446060.1 | - | 2 | 173 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI827524.1 | - | 2 | 128 | Chr. 13 - 99196953 - 99200710 [-] on Build GRCh38 |
