siRNA Details
Gene Symbol: LOC101927021 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110065.1 | 4 | 810 | |||
NR_110066.1 | 3 | 492 | ||||
NR_110067.1 | 4 | 748 | ||||
NR_110068.1 | 2 | 448 | ||||
NR_110069.1 | 3 | 640 | ||||
NR_110070.1 | 5 | 599 | ||||
GenBank mRNA | AK090593.1 | - | 3 | 374 | ||
DA025114.1 | - | 3 | 374 | |||
DA689880.1 | - | 2 | 328 | |||
HY003139.1 | - | 3 | 464 |
