siRNA Details
Gene Symbol: ASTN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033973.1 | 6 | 882 | |||
GenBank mRNA | AK057447.1 | - | 6 | 882 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ASTN2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033973.1 | 6 | 882 | |||
GenBank mRNA | AK057447.1 | - | 6 | 882 |