siRNA Details
Gene Symbol: VIM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108060.1 | 2 | 63 | |||
GenBank mRNA | BG492614.1 | - | 2 | 63 | ||
BG495699.1 | - | 2 | 63 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: VIM-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108060.1 | 2 | 63 | |||
GenBank mRNA | BG492614.1 | - | 2 | 63 | ||
BG495699.1 | - | 2 | 63 |