siRNA Details
Gene Symbol: EPHA1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033897.1 | 5 | 4179 | |||
GenBank mRNA | AL833583.1 | - | 5 | 4180 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG621177.1 | - | 1 | 135 | Chr. 7 - 143522476 - 143523174 [+] on Build GRCh38 |
