siRNA Details
Gene Symbol: LINC00710 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015413.1 | 2 | 284 | |||
NR_024410.1 | 2 | 284 | ||||
GenBank mRNA | BC033338.1 | - | 2 | 216 | ||
BC106956.2 | - | 2 | 88 | |||
DB038073.1 | - | 2 | 284 |
