siRNA Details
Gene Symbol: LOC100996624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132438.1 | 2 | 1052 | |||
GenBank mRNA | BC037254.1 | - | 2 | 1052 | ||
BC045661.1 | - | 2 | 991 | |||
BC047650.1 | - | 2 | 991 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100996624 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132438.1 | 2 | 1052 | |||
GenBank mRNA | BC037254.1 | - | 2 | 1052 | ||
BC045661.1 | - | 2 | 991 | |||
BC047650.1 | - | 2 | 991 |