siRNA Details
Gene Symbol: MCHR2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038384.1 | 3 | 511 | |||
GenBank mRNA | BU853836.1 | - | 3 | 484 | ||
BX097456.1 | - | 2 | 336 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: MCHR2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038384.1 | 3 | 511 | |||
GenBank mRNA | BU853836.1 | - | 3 | 484 | ||
BX097456.1 | - | 2 | 336 |