siRNA Details
Gene Symbol: LINC00358 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046995.1 | 1 | 126 | |||
GenBank mRNA | BC037867.2 | - | 1 | 126 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI464936.1 | - | 1 | 131 | Chr. 13 - 62004093 - 62029548 [-] on Build GRCh38 |
