siRNA Details
Gene Symbol: LINC01501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034157.1 | 6 | 680 | |||
GenBank mRNA | BC040625.1 | - | 6 | 680 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX118902.1 | - | 3 | 214 | Chr. 9 - 90463224 - 90505759 [-] on Build GRCh38 |
