siRNA Details
Gene Symbol: ISPD-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038946.1 | 1 | 80 | |||
NR_038947.1 | 1 | 80 | ||||
GenBank mRNA | BC034288.1 | - | 1 | 73 | ||
BC047638.1 | - | 1 | 73 | |||
BX115602.1 | - | 1 | 22 | |||
DB462496.1 | - | 1 | 83 |
