siRNA Details
Gene Symbol: ISPD-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038946.1 | 5 | 856 | |||
NR_038947.1 | 5 | 675 | ||||
GenBank mRNA | BC034288.1 | - | 6 | 849 | ||
BC047638.1 | - | 6 | 849 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: ISPD-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038946.1 | 5 | 856 | |||
NR_038947.1 | 5 | 675 | ||||
GenBank mRNA | BC034288.1 | - | 6 | 849 | ||
BC047638.1 | - | 6 | 849 |