siRNA Details
Gene Symbol: STEAP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046721.1 | 2 | 1234 | |||
GenBank mRNA | AK127773.1 | - | 2 | 1233 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: STEAP3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046721.1 | 2 | 1234 | |||
GenBank mRNA | AK127773.1 | - | 2 | 1233 |