siRNA Details
Gene Symbol: ZRANB2-AS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046217.1 | 4 | 307 | |||
GenBank mRNA | BM766420.1 | - | 4 | 309 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU561405.1 | - | 4 | 266 | Chr. 1 - 71081365 - 71237715 [+] on Build GRCh38 |
