siRNA Details
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 5 | 1032 | |||
NR_027429.1 | 4 | 945 | ||||
NR_027430.1 | 4 | 821 | ||||
NR_027431.2 | 4 | 795 | ||||
NR_027432.1 | 3 | 708 | ||||
GenBank mRNA | AK021673.1 | - | 4 | 664 | ||
AK096758.1 | - | 5 | 734 | |||
AK308811.1 | - | 3 | 261 | |||
AK309654.1 | - | 4 | 444 | |||
AK310182.1 | - | 4 | 497 | |||
AK310261.1 | - | 3 | 398 | |||
BC028046.1 | - | 3 | 369 |
