siRNA Details
Gene Symbol: FLJ33581 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040102.1 | 2 | 293 | |||
GenBank mRNA | AK090900.1 | - | 2 | 293 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA153363.1 | - | 2 | 293 | Chr. 20 - 24199767 - 24211844 [+] on Build GRCh38 |
