siRNA Details
Gene Symbol: MIRLET7DHG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046163.1 | 4 | 1231 | |||
NR_046165.1 | 3 | 1172 | ||||
GenBank mRNA | BC036695.1 | - | 4 | 1210 | ||
BC045813.1 | - | 3 | 1172 | |||
BX088742.1 | - | 1 | 135 |
