siRNA Details
Gene Symbol: DSCR9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026719.2 | 3 | 281 | |||
GenBank mRNA | AB066100.1 | - | 1 | 48 | ||
AB212286.1 | - | 3 | 281 | |||
AB212287.1 | - | 4 | 339 | |||
AB212288.1 | - | 5 | 428 | |||
AB212290.1 | - | 2 | 239 | |||
AB212291.1 | - | 4 | 370 | |||
BC066653.1 | - | 3 | 281 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ223675.1 | - | 280 | Chr. - - [-] on Build |
