siRNA Details
Gene Symbol: PSMG3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021487.2 | 6 | 3809 | |||
NR_027328.1 | 4 | 5650 | ||||
NR_027329.1 | 5 | 5758 | ||||
GenBank mRNA | AB067495.1 | - | 4 | 5628 | ||
AK091044.1 | - | 1 | 3005 | |||
AK092212.1 | - | 1 | 2252 | |||
BC036405.1 | - | 6 | 3617 | |||
BC047649.1 | - | 6 | 3612 |
