siRNA Details
Gene Symbol: PSMG3-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021487.2 | 2 | 513 | |||
NR_027328.1 | 2 | 259 | ||||
NR_027329.1 | 2 | 259 | ||||
GenBank mRNA | AB067495.1 | - | 2 | 237 | ||
AK054665.1 | - | 2 | 259 | |||
BC036405.1 | - | 2 | 321 | |||
BC047649.1 | - | 2 | 316 | |||
BX538028.1 | - | 2 | 197 | |||
DA046883.1 | - | 2 | 259 |
