siRNA Details
Gene Symbol: LIMD1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033947.1 | 2 | 636 | |||
GenBank mRNA | AK026206.1 | - | 2 | 637 | ||
BC047037.1 | - | 2 | 244 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LIMD1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_033947.1 | 2 | 636 | |||
GenBank mRNA | AK026206.1 | - | 2 | 637 | ||
BC047037.1 | - | 2 | 244 |