siRNA Details
Gene Symbol: ZNF341-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110623.1 | 1, 2 | 52 | |||
GenBank mRNA | AA453965.1 | - | 1, 2 | 52 | ||
BX102976.1 | - | 1, 2 | 50 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY003473.1 | - | 1, 2 | 25 | Chr. 20 - 33787480 - 33811071 [-] on Build GRCh38 |
