siRNA Details
Gene Symbol: HCG18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024052.2 | 5 | 2645 | |||
NR_024053.2 | 4 | 2431 | ||||
NR_102326.1 | 3 | 4302 | ||||
GenBank mRNA | AK055657.1 | - | 5 | 2339 | ||
AK056160.1 | - | 4 | 1648 | |||
AK098416.1 | - | 2 | 1181 |
