siRNA Details
Gene Symbol: LOC101926941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120664.1 | 3 | 361 | |||
GenBank mRNA | AI700977.1 | - | 3 | 356 | ||
DA537977.1 | - | 3 | 361 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101926941 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120664.1 | 3 | 361 | |||
GenBank mRNA | AI700977.1 | - | 3 | 356 | ||
DA537977.1 | - | 3 | 361 |