siRNA Details
Gene Symbol: GS1-600G8.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046087.1 | 16 | 1607 | |||
GenBank mRNA | BC022040.1 | - | 16 | 1612 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GS1-600G8.3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046087.1 | 16 | 1607 | |||
GenBank mRNA | BC022040.1 | - | 16 | 1612 |