siRNA Details
Gene Symbol: GNA14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121184.1 | 2 | 234 | |||
GenBank mRNA | BC043514.2 | - | 1 | 200 | ||
BI830360.1 | - | 2 | 234 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GNA14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121184.1 | 2 | 234 | |||
GenBank mRNA | BC043514.2 | - | 1 | 200 | ||
BI830360.1 | - | 2 | 234 |