siRNA Details
Gene Symbol: LOC101928650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096329.1 | - | 3 | 795 | ||
BC065239.1 | - | 2 | 895 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928650 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK096329.1 | - | 3 | 795 | ||
BC065239.1 | - | 2 | 895 |