siRNA Details
Gene Symbol: LINC01445 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120515.1 | 3 | 885 | |||
NR_120516.1 | 3 | 418 | ||||
GenBank mRNA | AV709400.1 | - | 3 | 347 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX111274.1 | - | 3 | 418 | Chr. 7 - 54330697 - 54349641 [+] on Build GRCh38 |
