siRNA Details
Gene Symbol: LOC643355 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270945.1 | NP_001257874.1 | 2 | 1046 | ||
XM_005271117.3 | XP_005271174.1 | 3 | 1135 | |||
GenBank mRNA | BC034233.1 | - | 2 | 1078 |
