siRNA Details
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024366.1 | 3 | 5723 | |||
GenBank mRNA | AK127516.1 | - | 1 | 4709 | ||
AL832752.1 | - | 3 | 5547 | |||
BX647840.1 | - | 2 | 5950 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM225A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024366.1 | 3 | 5723 | |||
GenBank mRNA | AK127516.1 | - | 1 | 4709 | ||
AL832752.1 | - | 3 | 5547 | |||
BX647840.1 | - | 2 | 5950 |