siRNA Details
Gene Symbol: DPP10-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132105.1 | 1 | 60 | |||
GenBank mRNA | BC033566.1 | - | 1 | 60 | ||
HY010208.1 | - | 1 | 40 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DPP10-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132105.1 | 1 | 60 | |||
GenBank mRNA | BC033566.1 | - | 1 | 60 | ||
HY010208.1 | - | 1 | 40 |