siRNA Details
Gene Symbol: LOC101928489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110889.1 | 2 | 1538 | |||
GenBank mRNA | AK124950.1 | - | 2 | 1427 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC101928489 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110889.1 | 2 | 1538 | |||
GenBank mRNA | AK124950.1 | - | 2 | 1427 |