siRNA Details
Gene Symbol: LINC01273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_109943.1 | 3 | 405 | |||
GenBank mRNA | BM750089.1 | - | 3 | 386 | ||
BQ646539.1 | - | 3 | 405 | |||
BX091132.1 | - | 2 | 161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM832820.1 | - | 3 | 387 | Chr. 20 - 50172590 - 50176499 [+] on Build GRCh38 |
