siRNA Details
Gene Symbol: TMEM44-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047573.1 | 3 | 394 | |||
NR_047574.1 | 3 | 390 | ||||
GenBank mRNA | BM717338.1 | - | 3 | 252 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG764808.1 | - | 2 | 165 | Chr. 3 - 194589070 - 194590259 [+] on Build GRCh38 |
