siRNA Details
Gene Symbol: LOC101926944 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110627.1 | 1 | 109 | |||
GenBank mRNA | BC042048.1 | - | 1 | 109 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BM556913.1 | - | 119 | Chr. - - [-] on Build |
