siRNA Details
Gene Symbol: IDH1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046452.1 | 2 | 352 | |||
GenBank mRNA | BJ994720.1 | - | 2 | 353 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: IDH1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046452.1 | 2 | 352 | |||
GenBank mRNA | BJ994720.1 | - | 2 | 353 |