siRNA Details
Gene Symbol: NEBL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046283.1 | 2 | 470 | |||
NR_046284.1 | 2 | 424 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: NEBL-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046283.1 | 2 | 470 | |||
NR_046284.1 | 2 | 424 |