siRNA Details
Gene Symbol: LEF1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029373.1 | 2, 3 | 1047 | |||
NR_029374.1 | 2, 3 | 417 | ||||
GenBank mRNA | BC020624.1 | - | 2, 3 | 417 | ||
BC041819.1 | - | 2, 3 | 1047 | |||
BC067877.1 | - | 2, 3 | 552 |
