siRNA Details
Gene Symbol: LOC284825 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126010.1 | 2 | 216 | |||
NR_126011.1 | 3 | 265 | ||||
GenBank mRNA | AK093119.1 | - | 2 | 216 | ||
BC039446.1 | - | 2 | 183 | |||
CD515480.1 | - | 3 | 265 |
