siRNA Details
Gene Symbol: LOC102724511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125878.1 | 2, 3 | 310 | |||
GenBank mRNA | BX092407.1 | - | 2, 3 | 280 | ||
HY033083.1 | - | 2, 3 | 310 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC102724511 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125878.1 | 2, 3 | 310 | |||
GenBank mRNA | BX092407.1 | - | 2, 3 | 280 | ||
HY033083.1 | - | 2, 3 | 310 |