siRNA Details
Gene Symbol: GNG12-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040077.1 | 10 | 1308 | |||
GenBank mRNA | AK124028.1 | - | 10 | 1293 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: GNG12-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040077.1 | 10 | 1308 | |||
GenBank mRNA | AK124028.1 | - | 10 | 1293 |