siRNA Details
Gene Symbol: DAOA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040247.1 | 7 | 1676 | |||
GenBank mRNA | AY138548.1 | - | 7 | 1676 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DAOA-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040247.1 | 7 | 1676 | |||
GenBank mRNA | AY138548.1 | - | 7 | 1676 |