siRNA Details
Gene Symbol: BPESC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026783.3 | 2, 3 | 2323 | |||
GenBank mRNA | AF196865.1 | - | 2, 3 | 2322 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: BPESC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026783.3 | 2, 3 | 2323 | |||
GenBank mRNA | AF196865.1 | - | 2, 3 | 2322 |