siRNA Details
Gene Symbol: LOC151484 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038238.1 | 3 | 729 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI821930.1 | - | 3 | 725 | Chr. 2 - 230886546 - 230896103 [+] on Build GRCh38 |
