siRNA Details
Gene Symbol: IDI2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024628.1 | 1 | 100 | |||
NR_024629.1 | 1 | 100 | ||||
NR_027708.1 | 1 | 100 | ||||
NR_027709.1 | 1 | 100 | ||||
GenBank mRNA | AF220183.1 | - | 1 | 71 | ||
AK314921.1 | - | 1 | 100 | |||
BF724542.1 | - | 1 | 63 |
