siRNA Details
Gene Symbol: IDI2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024628.1 | 4 | 821 | |||
NR_024629.1 | 3 | 658 | ||||
NR_027708.1 | 5 | 954 | ||||
NR_027709.1 | 5 | 1038 | ||||
GenBank mRNA | AF220183.1 | - | 3 | 629 | ||
AK314921.1 | - | 3 | 658 | |||
BC104155.1 | - | 4 | 711 | |||
BC104156.1 | - | 4 | 711 | |||
BC112975.1 | - | 4 | 711 | |||
BC112976.1 | - | 4 | 711 |
