siRNA Details
Gene Symbol: LOC100507670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135595.1 | 1 | 161 | |||
GenBank mRNA | BC021024.1 | - | 1 | 161 | ||
BE385446.1 | - | 1 | 189 | |||
DA674483.1 | - | 1 | 129 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100507670 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135595.1 | 1 | 161 | |||
GenBank mRNA | BC021024.1 | - | 1 | 161 | ||
BE385446.1 | - | 1 | 189 | |||
DA674483.1 | - | 1 | 129 |