siRNA Details
Gene Symbol: KIZ-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042893.1 | - | 4 | 1585 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB085029.1 | - | 6 | 515 | Chr. 20 - 21197462 - 21218289 [-] on Build GRCh38 |
