siRNA Details
Gene Symbol: SLCO4A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024470.1 | 4 | 1123 | |||
GenBank mRNA | BC025345.1 | - | 4 | 1123 | 4862156 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SLCO4A1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024470.1 | 4 | 1123 | |||
GenBank mRNA | BC025345.1 | - | 4 | 1123 | 4862156 |